Learn More
Aviva Systems Biology Corp PTRH2 Antibody - C-terminal region (ARP44695_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44695_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against PTRH2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human PTRH2. Alternate names: BIT1, CGI-147, FLJ32471, PTH2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-53431