Learn More
ATCC pTR-UF-11 plasmid; in E. coli
Catalog No. 502384252
Encompass
pTR-UF11 is an adeno-associated virus-based plasmid vector containing a CMV IE enhancer and chicken Beta-actin promoter driving the hGFP gene. It is distributed in Escherichia coli.
Catalog No. 50-238-4252 Supplier ATCC Supplier No. MBA331
pTR-UF11 is an adeno-associated virus-based plasmid vector containing a CMV IE enhancer and chicken Beta-actin promoter driving the hGFP gene. It is distributed in Escherichia coli.