Learn More
PTS Diagnostics PTS Panels™ CHOL+HDL+GLU Panel Test Strips
Description
Includes
Vial of test strips and desiccant, MEMo Chip (contains lot-specific test strip information), Instructions
Requires
- (Not provided)
- CardioChek PA analyzer, Quality Control Materials, Lancets for fingerstick (or venous blood collection supplies), Alcohol wipes and/or gauze, Capillary Blood Collector or other precision pipet for blood collection and application
- Accurate total cholesterol, HDL Cholesterol and glucose results in as little as 90 seconds.
- For use with the CardioChek Plus analyzer and 40 μL PTS Collect capillary tubes. (Each sold separately.)
- Can be stored at room temperature.
- Includes one 15-count vial of CHOL+HDL+GLU test strips and one MEMo Chip.
- 15 tests per box.
- 288 Box/Cs.
Specifications
Specifications
| Color Code | Black/Orange |
| Content And Storage | Room Temperature |
| Clia Complexity | Waived |
| Detectable Analytes | Total Cholesterol, HDL Cholesterol |
| Type | Test Strip |
| Disposable | Single-use |
| For Use With (Application) | In vitro diagnostic use in a professional healthcare setting |
| Includes | Lot-specific MEMo Chip |
| Quantity | 15 Pack |
| Range | Total Cholesterol: 100 to 400mg/dL (2.59 to 10.36mM/L), HDL Cholesterol: 15 to 100mg/dL (0.39 to 2.59mM/L), Glucose: 20 to 600mg/dL (1.11 to 3.33mM/L) |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.