Learn More
Medchemexpress LLC Ptupb | 1287761-01-6 | 98.1% | 100 MG
SDP
Catalog No. 5000230838
Encompass_Preferred
PTUPB is a small-molecule research compound that acts as a dual inhibitor of soluble epoxide hydrolase (sEH) and cyclooxygenase-2 (COX-2), used in preclinical studies to probe combined modulation of these pathways.
  • Potent dual inhibitor of sEH and COX-2 (sEH IC50 0.9 nM; COX-2 IC50 1.26 μM).
  • High purity, suitable for research applications.
  • Solid form with a molecular weight of 543.56 g/mol and formula C26H24F3N5O3S.
  • Stable as a powder at -20°C; in solvent, long-term storage recommended at -80°C.
  • Intended for research use only; not for human or diagnostic use.
Catalog No. 50-002-30838 Supplier Medchemexpress LLC Supplier No. HY122591100MG
PTUPB is a small-molecule research compound that acts as a dual inhibitor of soluble epoxide hydrolase (sEH) and cyclooxygenase-2 (COX-2), used in preclinical studies to probe combined modulation of these pathways.
  • Potent dual inhibitor of sEH and COX-2 (sEH IC50 0.9 nM; COX-2 IC50 1.26 μM).
  • High purity, suitable for research applications.
  • Solid form with a molecular weight of 543.56 g/mol and formula C26H24F3N5O3S.
  • Stable as a powder at -20°C; in solvent, long-term storage recommended at -80°C.
  • Intended for research use only; not for human or diagnostic use.