Learn More
New England Biolabs, Inc. pTWIN1 Vector DNA – 10 µg
SDP
Catalog No. NC0832508
Encompass
pTWIN1 is an E. coli expression vector designed for protein purification or for the isolation of proteins with an N-terminal cysteine and/or a C-terminal thioester. A polylinker in the vector is designed for the in-frame fusion of a target gene between the modified Ssp DnaB and Mxe GyrA inteins. The presence of the chitin binding domain from Bacillus circulans facilitates purification.
Catalog No. NC0832508 Supplier New England Biolabs, Inc. Supplier No. N6951S
pTWIN1 is an E. coli expression vector designed for protein purification or for the isolation of proteins with an N-terminal cysteine and/or a C-terminal thioester. A polylinker in the vector is designed for the in-frame fusion of a target gene between the modified Ssp DnaB and Mxe GyrA inteins. The presence of the chitin binding domain from Bacillus circulans facilitates purification.