Learn More
Biomatik Corporation Ptx3 Polyclonal Antibody
Supplier:  Biomatik Corporation CAC11201
Encompass_Preferred
Ptx3 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to PTX3. The Ptx3 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Pentraxin-Related Protein Ptx3 Protein (99-164Aa). Applications: ELISA, WB, IHC, IF. Target Synonyms: PTX3. Accession Number: P26022. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4564