Learn More
Enzo Life Sciences PTX3 monoclonal antibody (MNB1) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX804463C100
Encompass_Preferred
Clone: MNB1. Isotype: IgG2b. Immunogen: Recombinant human PTX3 (pentraxin 3). Species reactivity: Human, Mouse. Specificity: Recognizes the C-terminus of PTX3. Applications: IHC (FS), IHC (PS), WB. Formulation: Liquid. Purified antibody in PBS containing 0.1% sodium azide.
Catalog No. 50-200-9915