Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents QP10121MA20UG
Encompass_Preferred
Product Type: Recombinant Protein. Conjugate/Tag: Flag . Host: Mammal. Species Reactivity: Bovine. Protein Expression: A DNA sequence encoding the Bos taurus (Bovine) PTX3 / Pentraxin 3 / TSG-14, was expressed in the hosts and tags indicated. AA Sequence: ENSDDYELMY VNLDNEIDNG LHPTEDPTPC DCSRENSEWD KLFTMLENSQ MREGMLLQAT DVMLRGELQK LQAELGRLEG SLQKLCGPEA PSETRLARAL DDLLQASRDA GRRLARLEDA GALRPQEEAG RALGAVLEEL RRTRADLRAV QGWAASRWLP AGCETAILFP MRSKKIFASV HPVTPMKLET FSACIWVKAT EVLNKTVLFS YGTKRNPYEI QLYLSYRSIM LVVGGEENRL VADAVISLGT WTHLCSTWDS KKGHMALWVN GDSVATAVDM ATGHVVPEGG ILQIGQEKNG CCVGGGFDET LAFSGRLTGF NIWEGVLSNE EIREAGGAES CHIRGNVVGW GVTEIQPHGG AQYVY. Buffer: Tris-based buffer, 50% glycerol. Purity: Greater than 90% as determined by SDS-PAGE. Endotoxin: Not determined. Gene Symbol: PTX3. Gene ID: 541148. Uniprot: Q0VCG9.
Catalog No. 50-188-5501