Learn More
New England Biolabs, Inc. pTYB21 Vector – 10 µg
SDP
Catalog No. 504423
Encompass
pTYB21 is an E. coli expression N-terminal fusion vector designed for in-frame insertion of a target gene into the polylinker downstream of the intein tag. This allows the N-terminus of the target protein to be fused to the intein tag. The self-cleavage activity of the intein allows the release of the target protein from the chitin-bound intein tag, resulting in a single column purification of the target protein.
Catalog No. 50-442-3 Supplier New England Biolabs, Inc. Supplier No. N6709S
pTYB21 is an E. coli expression N-terminal fusion vector designed for in-frame insertion of a target gene into the polylinker downstream of the intein tag. This allows the N-terminus of the target protein to be fused to the intein tag. The self-cleavage activity of the intein allows the release of the target protein from the chitin-bound intein tag, resulting in a single column purification of the target protein.