Learn More
Enzo Life Sciences [pTyr1146] Insulin receptor (1142-1153) (0.5mg)
SDP
Supplier:  Enzo Life Sciences BMLP3150500
Encompass_Preferred
This phosphopeptide sequence, TRDIpYETDYYRK, corresponds to the insulin receptor regulatory region. Can be used as a tyrosine phosphatase substrate. Purity: ≥95% (HPLC). Formulation: Lyophilized. Long Term Storage: -20°C.
Catalog No. 50-201-1342