Learn More
Enzo Life Sciences [pTyr1150] Insulin receptor (1142-1153) (0.5mg)
SDP
Supplier:  Enzo Life Sciences BMLP3160500
Encompass_Preferred
This phosphopeptide, TRDIYETDpYYRK, corresponds to the insulin receptor regulatory region. Can be used as a tyrosine phosphatase substrate. Long Term Storage: -20°C.
Catalog No. 50-201-1343