Learn More
Medchemexpress LLC HY-103118 10mg Medchemexpress, PU02 CAS:313984-77-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10311810MG
Encompass_Preferred
Medchemexpress, HY-103118 10mg PU02 CAS:313984-77-9 PU02, a derivative of 6-MP (HY-13677), is a negative allosteric modulator (NAM) of 5-HT3 receptor, with IC50 values of 0.36 and 0.73 μM in HEK293 cells transfected with human 5-HT3A and 5-HT3AB receptors respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0657