Learn More
New England Biolabs, Inc. pUC19 Vector – 50 µg
SDP
Catalog No. 50811771
Encompass
pUC19 is a commonly used cloning vector that conveys the Amp resistance. The molecule is a small double-stranded circle, 2686 base pairs in length, and has a high copy number. pUC19 carries a 54 base-pair multiple cloning site polylinker that contains unique sites for 13 different hexanucleotide-specific restriction endonucleases.
Catalog No. 50-811-771 Supplier New England Biolabs, Inc. Supplier No. N3041S
pUC19 is a commonly used cloning vector that conveys the Amp resistance. The molecule is a small double-stranded circle, 2686 base pairs in length, and has a high copy number. pUC19 carries a 54 base-pair multiple cloning site polylinker that contains unique sites for 13 different hexanucleotide-specific restriction endonucleases.