Learn More
Genscript Corporation pUC57 plasmid DNA
Catalog No. 5000057479
Encompass
pUC57 is a common used plasmid cloning vector in i E coli /i The vector length is 2 710 bp and is isolated from i E coli /i strain DH5 945 by standard procedures br b Note /b Supplied in lyophilized form
Catalog No. 50-000-57479 Supplier Genscript Corporation Supplier No. SD1176
pUC57 is a common used plasmid cloning vector in i E coli /i The vector length is 2 710 bp and is isolated from i E coli /i strain DH5 945 by standard procedures br b Note /b Supplied in lyophilized form