Learn More
Mitegen Puck Dewar Loading Tool, For Separating & Loading Uni-Pucks into ALS-Robot Dewars, 2-Piece Design
SDP
Catalog No. 501187182
Encompass
This Puck Dewar Loading Tool is an indispensable accessory designed to streamline the precise and safe transfer of pucks or Uni-Pucks into an ALS-robot dewar. Engineered for accuracy and ease of use, this specialized tool consists of two distinct components: a durable Teflon tube part and a robust metal rod part. Together, these pieces facilitate the gentle separation and controlled placement of pucks, minimizing direct handling and reducing the risk of thermal shock or accidental contamination of cryogenic samples. The design ensures seamless integration into automated laboratory workflows, enhancing efficiency during critical sample loading procedures. By enabling precise manipulation of pucks, this tool safeguards valuable research materials and contributes to consistent, reliable results in high-throughput cryo-electron microscopy and crystallography applications, ensuring optimal cryogenic sample management.
Catalog No. 50-118-7182 Supplier Mitegen Supplier No. MCP111027
This Puck Dewar Loading Tool is an indispensable accessory designed to streamline the precise and safe transfer of pucks or Uni-Pucks into an ALS-robot dewar. Engineered for accuracy and ease of use, this specialized tool consists of two distinct components: a durable Teflon tube part and a robust metal rod part. Together, these pieces facilitate the gentle separation and controlled placement of pucks, minimizing direct handling and reducing the risk of thermal shock or accidental contamination of cryogenic samples. The design ensures seamless integration into automated laboratory workflows, enhancing efficiency during critical sample loading procedures. By enabling precise manipulation of pucks, this tool safeguards valuable research materials and contributes to consistent, reliable results in high-throughput cryo-electron microscopy and crystallography applications, ensuring optimal cryogenic sample management.