Learn More
Mitegen Uni-Puck Puck Separator Tools Set, Includes Base Removal Platform & Porcupine Push Tool, For Efficient Uni-Puck Separation
SDP
Supplier:  Mitegen MCP111028
Encompass
This Puck Separator Tools Set provides a specialized solution for efficiently and safely separating pucks or Uni-Pucks in cryogenic sample handling workflows. This essential set includes two distinct components: a robust base removal platform and a precision-engineered porcupine push tool. The base removal platform provides a stable surface for disassembling pucks, while the porcupine push tool allows for gentle yet firm pressure to separate puck components without causing damage to the pucks or valuable samples. Designed for ease of use, these tools minimize direct handling of cold samples and reduce the risk of thermal shock or contamination during critical cryogenic procedures. This set enhances laboratory efficiency and ensures the integrity of cryo-preserved specimens by streamlining the process of preparing and managing pucks for storage or experimental setups.
Catalog No. 50-118-7183