Learn More
Molecular Dimensions PUCK SEPARATOR TOOLS
SDP
Catalog No. 501038610
Encompass
Puck Separator Tools This set of tools includes a base removal platform and a porcupine push tool to recuperate your samples.
Catalog No. 50-103-8610 Supplier Molecular Dimensions Supplier No. MD7605
Puck Separator Tools This set of tools includes a base removal platform and a porcupine push tool to recuperate your samples.