Learn More
Molecular Dimensions PUCK-SHELVED SHIPPING CANE
SDP
Catalog No. 501038608
Encompass
Puck-Shelved Shipping Cane This set includes a shelved shipping cane, locking rod and hooked handle. Seven Uni-pucks may be shipped or stored in this cane. This fits into the CX100 transport Dewar (Dry Shipper - MD7-21).
Catalog No. 50-103-8608 Supplier Molecular Dimensions Supplier No. MD7603
Puck-Shelved Shipping Cane This set includes a shelved shipping cane, locking rod and hooked handle. Seven Uni-pucks may be shipped or stored in this cane. This fits into the CX100 transport Dewar (Dry Shipper - MD7-21).