Learn More
Mitegen Puck Wand For UniPucks
SDP
Catalog No. 501187181
Encompass
UniPuck tool with magnetic flat base, useful for handling UniPuck enclosures. Has insulated handling to help reduce cooling while handling pucks in cryogenic environment.
Catalog No. 50-118-7181 Supplier Mitegen Supplier No. MCP111026
UniPuck tool with magnetic flat base, useful for handling UniPuck enclosures. Has insulated handling to help reduce cooling while handling pucks in cryogenic environment.