Learn More
Apexbio Technology LLC Puerarin 3681-99-0 10mg
SDP
Catalog No. 5000097059
Encompass_Preferred
Puerarin (CAS 3681-99-0) is an isoflavone compound isolated from the root of Pueraria lobata It exerts multiple pharmacological effects primarily through modulation of oxidative stress and inflammatory signaling pathways Puerarin has been reported to enhance myocardial perfusion protect against ischemic injury and regulate glucose and lipid metabolism in preclinical models Additionally it demonstrates neuroprotective properties including mitigation of neuronal damage associated with neurodegenerative conditions Puerarin is widely studied for its potential in cardiovascular metabolic and neurological disease research
Catalog No. 50-000-97059 Supplier Apexbio Technology LLC Supplier No. N111410
Puerarin (CAS 3681-99-0) is an isoflavone compound isolated from the root of Pueraria lobata It exerts multiple pharmacological effects primarily through modulation of oxidative stress and inflammatory signaling pathways Puerarin has been reported to enhance myocardial perfusion protect against ischemic injury and regulate glucose and lipid metabolism in preclinical models Additionally it demonstrates neuroprotective properties including mitigation of neuronal damage associated with neurodegenerative conditions Puerarin is widely studied for its potential in cardiovascular metabolic and neurological disease research