Learn More
Medchemexpress LLC Puerarin | 3681-99-0 | 98.9% | 416.38 | 1 ML
SDP
Catalog No. 5000428087
Encompass_Preferred
Puerarin is an isoflavone extracted from Pueraria root, functioning as a 5-HT2C receptor antagonist. It inhibits the dorsal motor nucleus of the vagus (DMV)-vagus nerve pathway, which leads to decreased fat absorption.
  • Acts as a 5-HT2C receptor antagonist
  • Inhibits LPS-induced iNOS, COX-2, and CRP protein and mRNA expression
  • Functions as a novel open-channel blocker of IK1
  • Alleviates hepatic damage from chronic alcohol administration
  • Improves early-stage renal damage in diabetic rats
Catalog No. 50-004-28087 Supplier Medchemexpress LLC Supplier No. HYN014510MM/1ML
Puerarin is an isoflavone extracted from Pueraria root, functioning as a 5-HT2C receptor antagonist. It inhibits the dorsal motor nucleus of the vagus (DMV)-vagus nerve pathway, which leads to decreased fat absorption.
  • Acts as a 5-HT2C receptor antagonist
  • Inhibits LPS-induced iNOS, COX-2, and CRP protein and mRNA expression
  • Functions as a novel open-channel blocker of IK1
  • Alleviates hepatic damage from chronic alcohol administration
  • Improves early-stage renal damage in diabetic rats