Learn More
TARGETMOL CHEMICALS INC PUERARIN 6-O-XYLOSIDE 5MG
SDP
Catalog No. 502579710
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Puerarin 6''-O-xyloside possesses significant anti-osteoporotic activity on ovariectomy mice it has significant antitumour activities the mechanisms are related to increased levels of caspase-3 caspase-7 caspase-9 and Bax and reduced levels Bcl-2. purity: 98%
Catalog No. 50-257-9710 Supplier TARGETMOL CHEMICALS INC Supplier No. T5S20835MG
Also available in 1 mg 10 mg 25 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Puerarin 6''-O-xyloside possesses significant anti-osteoporotic activity on ovariectomy mice it has significant antitumour activities the mechanisms are related to increased levels of caspase-3 caspase-7 caspase-9 and Bax and reduced levels Bcl-2. purity: 98%