Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Puerarin, 98%

Catalog No. p-7043246
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6652803 1 g
AAJ6652806 5 g
AAJ6652809 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ6652803 Supplier Thermo Scientific Chemicals Supplier No. J6652803
Only null left
Add to Cart

CAS: 3681-99-0 | C21H20O9 | 416.38 g/mol

Thought to interfere with the synthesis of mycolic acids, an essential fatty acid component found in Mycobacterium cell walls

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Thought to interfere with the synthesis of mycolic acids, an essential fatty acid component found in Mycobacterium cell walls
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3681-99-0
Molecular Formula C21H20O9
Molecular Weight (g/mol) 416.38
MDL Number MFCD00063399
InChI Key HKEAFJYKMMKDOR-VPRICQMDSA-N
Synonym 8-beta-d-glucopyranosyl-4',7-dihydroxyisoflavone
PubChem CID 53384442
IUPAC Name 7-hydroxy-3-(4-hydroxyphenyl)-8-[(2S,3R,4R,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]-4H-chromen-4-one
SMILES OC[C@H]1O[C@H]([C@H](O)[C@@H](O)[C@@H]1O)C1=C2OC=C(C(=O)C2=CC=C1O)C1=CC=C(O)C=C1

Specifications

Melting Point 187°C to 189°C
Color White
Quantity 1 g
Beilstein 64198
Solubility Information Soluble in DMSO or DMF; Slightly soluble in water or ethanol
Formula Weight 416.38
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Puerarin
RTECSNumber UO5216000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.