Learn More
Medchemexpress LLC Puerarin apioside | 103654-50-8 | 99.7% | 548.49 | C26H28O13 | 10 MG
SDP
Catalog No. 5000295934
Encompass_Preferred
Mirificin (puerarin apioside) is an isoflavone isolated from Puerariae lobatae radix and used as a research reagent for biochemical and enzymatic studies. It has been reported to inhibit tyrosinase (TYR) with an IC50 of 12.66 μM and is provided as a high-purity solid suitable for assay development and natural product research.
  • High-purity compound for research use.
  • Inhibits tyrosinase; IC50 = 12.66 μM.
  • Chemical formula C26H28O13; molecular weight 548.49.
  • Suitable for enzyme inhibition and natural product characterization studies.
Catalog No. 50-002-95934 Supplier Medchemexpress LLC Supplier No. HYN213410MG
Mirificin (puerarin apioside) is an isoflavone isolated from Puerariae lobatae radix and used as a research reagent for biochemical and enzymatic studies. It has been reported to inhibit tyrosinase (TYR) with an IC50 of 12.66 μM and is provided as a high-purity solid suitable for assay development and natural product research.
  • High-purity compound for research use.
  • Inhibits tyrosinase; IC50 = 12.66 μM.
  • Chemical formula C26H28O13; molecular weight 548.49.
  • Suitable for enzyme inhibition and natural product characterization studies.