Learn More
Aviva Systems Biology Corp PUF60 Antibody - C-terminal region (ARP40760_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40760_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PUF60. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human PUF60. Alternate names: FIR, FLJ31379, RoBPI, SIAHBP1ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat, Zebrafish
Catalog No. 50-014-53234