Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PUFA No.1, Marine Source, Analytical Standard, MilliporeSigma™ Supelco™

Catalog No. 111003686 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-100-3686 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-100-3686 Supplier MilliporeSigma Supelco Supplier No. 47033
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PUFA No.1 is a complex qualitative standard mixture. Because it is extracted from natural materials, relative peak size and composition may vary from lot to lot.

For qualitative identification only. Product is extracted from natural sources. The fatty acid composition varies from lot-to-lot.
TRUSTED_SUSTAINABILITY

Specifications

Physical Form Neat
Synonym Polyunsaturated Fatty Acid Mix No0.1
Quantity 100 mg
Grade Analytical Standard
Concentration or Composition (by Analyte or Components) cis-4,7,10,13,16,19-Docosahexaenoic acid methyl ester; 11-Docosenoic acid methyl ester; Methyl all-cis-7,10,13,16,19-docosapentaenoate; Methyl cis-13-docosenoate; Methyl all-cis-5,8,11,14,17-eicosapentaenoate; Methyl linoleate; Methyl myristate; Methyl oleate; Methyl palmitate; Methyl palmitoleate; Methyl stearidonate; cis-11-Octadecenoic methyl ester; Methyl cis-11-eicosenoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.