Learn More
TARGETMOL CHEMICALS INC PULEGONE 50MG
SDP
Catalog No. 502576184
Encompass_Preferred
Also available in 5 mg 10 mg 25 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. 1. Pulegone ((+)-Pulegone) has cytotoxicity followed by regenerative cell proliferation is the MOA for Pulegone-induced urothelial tumors in female rats. 2. Pulegone induces a verapamil-sensitive psychostimulant effect that appears to independ on the opening of L-type calcium channels. 3. Pulegone has negative reinforcing properties and seems to possess anxiolytic-like actions unrelated to the benzodiazepine site of the (gamma)-aminobutyric acid type A (GABAA) receptor. purity: 98%
Catalog No. 50-257-6184 Supplier TARGETMOL CHEMICALS INC Supplier No. TCS010250MG
Add to Cart
Add to Cart
Also available in 5 mg 10 mg 25 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. 1. Pulegone ((+)-Pulegone) has cytotoxicity followed by regenerative cell proliferation is the MOA for Pulegone-induced urothelial tumors in female rats. 2. Pulegone induces a verapamil-sensitive psychostimulant effect that appears to independ on the opening of L-type calcium channels. 3. Pulegone has negative reinforcing properties and seems to possess anxiolytic-like actions unrelated to the benzodiazepine site of the (gamma)-aminobutyric acid type A (GABAA) receptor. purity: 98%