Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich ™-(+)-Pulegone

Catalog No. P55708100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P55708100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. P55708100ML Supplier Sigma Aldrich Supplier No. P55708100ML
Only null left
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(R)-(+)-Pulegone, a monoterpene ketone found in essential oil of pennyroyal, can induce abortion.(R)-(+)-Pulegone may be used as a starting material to synthesize:(2S,4R,6R,8S)-2,4,8-trimethyl-1,7-dioxaspiro[5.5]undecane, a spiroketal(R)-(+)-citronellic acid, an intermediate to prepare leucine-d3(+)-fawcettidine, a Lycopodium alkaloid

Specifications

CAS 20902-45-8
Melting Point 204°C
Molecular Formula C10H20N2O4S2
Linear Formula [-SC(CH3)2CH(NH2)CO2H]2
Refractive Index n20/D 1.486
MDL Number MFCD00004264
Quantity 100 mL
Synonym (+)-Pulegone; (R)-2-Isopropylidene-5-methylcyclohexanone; (R)-p-Menth-4(8)-en-3-one; p-Menth-4(8)-en-3-one
Molecular Weight (g/mol) 296.41 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.