Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(+)-Pulegone 95.0+%, TCI America™
SDP

Catalog No. M040710ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M040710ML 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M040710ML Supplier TCI America Supplier No. M040710ML

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 89-82-7
Molecular Formula C10H16O
Molecular Weight (g/mol) 152.237
MDL Number MFCD00063000
InChI Key NZGWDASTMWDZIW-QMMMGPOBSA-N
Synonym --pulegone, s---pulegone, pulegone,-, unii-hl7z89m60h, cyclohexanone, 5-methyl-2-1-methylethylidene-, s, s-p-menth-4 8-en-3-one, s-2-isopropylidene-5-methylcyclohexanone, 5s-5-methyl-2-propan-2-ylidenecyclohexan-1-one
PubChem CID 638012
ChEBI CHEBI:81226
IUPAC Name (5S)-5-methyl-2-propan-2-ylidenecyclohexan-1-one
SMILES CC1CCC(=C(C)C)C(=O)C1

Specifications

Color Yellow
Boiling Point 217°C
Quantity 10 mL
Formula Weight 152.24
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material (+)-Pulegone
EINECSNumber (3)-2400
RTECSNumber OT0261000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.