Learn More
ARBOR SCIENTIFIC PULL-BACK CAR
SDP
Catalog No. NC0382017
Encompass
Simply pull back on the spring-loaded, die-cast truck then release. Use a motion sensor or tape timer to measure the acceleration. The more you pull it backwards, the greater its acceleration. 5 in. (13cm) long.
Catalog No. NC0382017 Supplier ARBOR SCIENTIFIC Supplier No. P41980
Simply pull back on the spring-loaded, die-cast truck then release. Use a motion sensor or tape timer to measure the acceleration. The more you pull it backwards, the greater its acceleration. 5 in. (13cm) long.