Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Pull-handle Butterfly Valve Lockout ABS Plastic

Catalog No. 19129951 Shop All Brady Corporation Products
Change view
Click to view available options
Color:
Red
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-129-951 Red
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-129-951 Supplier Brady™ Supplier No. 170220
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Stop using ill-fitting, incorrect or unwieldy two-piece devices to lock out your pull handle valves. Get a safer solution that fits better and stays in place.

  • Exclusive, anti-pivot lockout device fits the full range of pull handle valve sizes, from ½ in. to 8 in. diameter
  • A convenient, one-piece design that stays together whether in use or in storage
  • Holds up to four worker padlocks
  • Effective for all popular pull handle valves commonly used in food and beverage, water utilities, chemical, pharmaceutical and many other industries

Specifications

Product Type Butterfly Valve Lockout
Color Red
Material ABS
Dimensions (L x W) 8.56 x 3.82 in.
For Use With (Application) Lockout Tagout
Height (English) 2.12 in.
Temperature Range (English) −40° to +100°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.