Learn More
Grainger Pull Station Hex Screw Double DPDT
Catalog No. 502766124
Encompass_Preferred
The Edwards Signaling 270-P double action fire alarm pull station is a manual fire alarm activation device designed for wall mounting. It features a dual-action mechanism that requires two motions to activate the alarm, helping prevent accidental activation. The pull station has a compact, low-profile design with a universal red finish and raised lettering. It is constructed of die-cast metal with a durable powder coat finish. The pull station is UL listed and meets ADA requirements. It requires a key to reset the alarm after activation. Overall, the 270-P provides a reliable and code-compliant manual activation device for fire alarm systems.
Catalog No. 50-276-6124 Supplier Grainger Supplier No. 16X415
The Edwards Signaling 270-P double action fire alarm pull station is a manual fire alarm activation device designed for wall mounting. It features a dual-action mechanism that requires two motions to activate the alarm, helping prevent accidental activation. The pull station has a compact, low-profile design with a universal red finish and raised lettering. It is constructed of die-cast metal with a durable powder coat finish. The pull station is UL listed and meets ADA requirements. It requires a key to reset the alarm after activation. Overall, the 270-P provides a reliable and code-compliant manual activation device for fire alarm systems.