Learn More
Grainger Pull Station Hex Screw Single DPDT
Catalog No. 502766690
Encompass_Preferred
The Edwards Signaling 120V AC Volt Pull Station is a manual fire alarm activation device designed for indoor use. It features a dual-action mechanism requiring two motions to activate the alarm, helping prevent accidental activation. The pull station has a compact, low-profile design with a universal pull lever that rotates down. It is constructed of Lexan with a red finish and includes an alarm-activating switch. The pull station operates on 120V AC power and provides a set of Form C contacts rated at 5 amps resistive at 125V AC. It meets ADA requirements. The device allows manual initiation of fire alarms in commercial buildings, schools, hospitals, and other facilities.
Catalog No. 50-276-6690 Supplier Grainger Supplier No. 16X413
The Edwards Signaling 120V AC Volt Pull Station is a manual fire alarm activation device designed for indoor use. It features a dual-action mechanism requiring two motions to activate the alarm, helping prevent accidental activation. The pull station has a compact, low-profile design with a universal pull lever that rotates down. It is constructed of Lexan with a red finish and includes an alarm-activating switch. The pull station operates on 120V AC power and provides a set of Form C contacts rated at 5 amps resistive at 125V AC. It meets ADA requirements. The device allows manual initiation of fire alarms in commercial buildings, schools, hospitals, and other facilities.