Learn More
Grainger Pull Station Key Reset Double DPDT
Catalog No. 502766204
Encompass_Preferred
The Edwards Signaling 270-P Fire Alarm Pull Station is a double pole, double throw (DPDT) manual fire alarm station. It features a dual-action mechanism that requires two motions to activate the alarm, helping prevent accidental activation. The pull station has a rugged die-cast metal housing finished in red with clearly visible operating instructions. It is compatible with multiple signaling systems. The pull station meets ADA requirements.
Catalog No. 50-276-6204 Supplier Grainger Supplier No. 16X414
The Edwards Signaling 270-P Fire Alarm Pull Station is a double pole, double throw (DPDT) manual fire alarm station. It features a dual-action mechanism that requires two motions to activate the alarm, helping prevent accidental activation. The pull station has a rugged die-cast metal housing finished in red with clearly visible operating instructions. It is compatible with multiple signaling systems. The pull station meets ADA requirements.