Learn More
Grainger Pull Station Reset Key
Catalog No. 502766206
Encompass_Preferred
The Edwards Signaling Pull Station Reset Key is a replacement key for Edwards pull stations. It is made of nickel plated brass and measures 1.25 inches long. The key has a round head with a diameter of 0.375 inches and is used to reset the pull station after the pull station has been activated. The key can be used with a variety of Edwards pull station models including the 270SPO-10, 270SPO-10RLY, 270SPO-12RLY, 270SPO-6RLY, and 270SPO-6RLY-IV. This is an OEM replacement part from Edwards Signaling.
Catalog No. 50-276-6206 Supplier Grainger Supplier No. 16X418
The Edwards Signaling Pull Station Reset Key is a replacement key for Edwards pull stations. It is made of nickel plated brass and measures 1.25 inches long. The key has a round head with a diameter of 0.375 inches and is used to reset the pull station after the pull station has been activated. The key can be used with a variety of Edwards pull station models including the 270SPO-10, 270SPO-10RLY, 270SPO-12RLY, 270SPO-6RLY, and 270SPO-6RLY-IV. This is an OEM replacement part from Edwards Signaling.