Learn More
Marketlab Pull Up™ Biopsy Probe Cover Set
Catalog No. 22505250
Change view
Click to view available options
Quantity:
24/Pk.
Catalog No. Quantity
22-505-250 24/Pk.
1 options
Catalog No. 22-505-250 Supplier Marketlab Supplier No. 934468
Add to Cart
Add to Cart
The larger sized Pull Up™ Biopsy Probe Cover accomodates brackets, guides and biopsy devices.
  • Set includes 5.5 x 48 in. sterile Pull Up™ Probe Covers, sterile gel, and bands for 24 procedures.
  • General purpose
  • Latex free
  • Cover with box-style 3D-end helps provide better probe coupling
Specifications
Quantity 24/Pk.