Learn More
Marketlab Pull Up™ Ultrasound Probe Cover Set with 3D End
Catalog No. 22505333
Change view
Click to view available options
Length (English):
48 in.
Catalog No. Length (English)
22-505-333 48 in.
1 options
Catalog No. 22-505-333 Supplier Marketlab Supplier No. 934430CO
Add to Cart
Add to Cart
Provides quick and easy loading of an ultrasound probe into a long, sterile probe cover.
  • Provides protection from cross contamination
  • Helps protect expensive ultrasound probes
  • Minimizes clean up by allowing the user to add gel only where necessary
  • Saves more time than with conventional covers that require assembly during procedure
  • General purpose, telescopic fold, and sterile
  • Includes sterile gel and bands
  • Cover with box-style 3D-end helps provide better probe coupling
  • Unique design allows the user to quickly find the open end of the probe cover.
  • Place the desired amount of gel inside, precisely position the probe into the end, and effortlessly pull the cover over the probe and probe cable.
  • Each package includes:
    • 1 Pull Up™ cover
    • 1 sterile gel pack
    • 2 elastic fasteners
Specifications
Quantity 24/Pk.
Length (English) 48 in.