Learn More
Narishige International Usa Inc Versatile and easy-to-use puller by automated double pulling
Catalog No. NC1402089
Encompass
PC-100 enables the user to produce many kinds of pipettes including pipettes with a tip of several 10µm to be used for the experiments in biotechnology, those with a tip of a few micrometers to be used for the experiments in electrophysiology, and even those with a tip in the region of 1µm. In terms of producing pipettes with a wide variety of shapes and tips, the function of Mode selector button allows to choose a pulling mode setting between one-stage and two-stage. Once the value has been determined, the specification of PC-100 allows the user to reproduce the same pipette as the last one. In addition, the PC-100 applies free fall by gravity as its pulling force (or descending by the action of a weight), thus it is provided with two types of weights; a light type and a heavy type, two pieces each that is attach/detachable respectively in order to adjust the pulling force.
Catalog No. NC1402089 Supplier Narishige International Usa Inc Supplier No. PC100
Add to Cart
Add to Cart
PC-100 enables the user to produce many kinds of pipettes including pipettes with a tip of several 10µm to be used for the experiments in biotechnology, those with a tip of a few micrometers to be used for the experiments in electrophysiology, and even those with a tip in the region of 1µm. In terms of producing pipettes with a wide variety of shapes and tips, the function of Mode selector button allows to choose a pulling mode setting between one-stage and two-stage. Once the value has been determined, the specification of PC-100 allows the user to reproduce the same pipette as the last one. In addition, the PC-100 applies free fall by gravity as its pulling force (or descending by the action of a weight), thus it is provided with two types of weights; a light type and a heavy type, two pieces each that is attach/detachable respectively in order to adjust the pulling force.