Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vernier™ Pulley Bracket
SDPSCI_ED

Catalog No. S16102ND Shop All Vernier Software Products
Change view
Click to view available options
Color: Black
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
S16102ND Color: Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S16102ND Supplier Vernier™ Supplier No. BSPA
Add to Cart
Add to Cart

Includes

Pulley Bracket

The Pulley Bracket attaches an Ultra Pulley to the end of a Vernier Dynamics Track.

  • The Pulley Bracket is used to attach a Vernier Ultra Pulley to a Track. The same Track is included with the Vernier Dynamics Cart and Track System.
  • Typically the Pulley Bracket is used when an experiment requires a string passing between a hanging mass and a dynamics cart. The configuration is commonly called a half-Atwood machine.
  • The Pulley Bracket can be assembled for use with or without a Vernier Photogate. When assembled with a photogate, the pulley spokes pass through the beam of the photogate, allowing the gate to perform motion timing.
  • To install the Pulley Bracket, loosen the wing bolt on the short leg of the bracket, and insert the oblong nut into the slot in the center underside of the track. Tighten the wing bolt with the bracket firmly against the end of the track. If a photogate will not be used in the setup, attach the pulley on the vertical arm of the bracket, away from the track, using the supplied round bolt. Do not include the black plastic bracket.
  • To add a photogate, slide the black plastic bracket over the metal bracket, with the open side facing away from the track and the bolt hole up. Place the photogate in the open slot and the pulley between the arms of the photogate.

Warranty and Services

5-year limited warranty

Specifications

Color Black
For Use With (Application) Educational use only
Includes Wing bolts
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.