Learn More
Medchemexpress LLC Pullulan | 9057-02-7 | MFCD00081940 | 98.0% | Mw 362-480 kDa; Mn 100-200 kDa | C20H36O16 | 10 G
SDP
Catalog No. 5000282898
Encompass_Preferred
Pullulan is a water-soluble linear glucosic polysaccharide produced by Aureobasidium pullulans. Supplied as a viscosity-specified powder standard for laboratory and formulation use, it offers defined molecular-weight distribution and high purity for food, pharmaceutical, and materials applications.
  • Provides controlled viscosity (10% solution: 15-180 mPa·s).
  • Defined molecular-weight distribution (Mw 362-480 kDa; Mn 100-200 kDa).
  • High purity suitable for analytical and formulation work.
  • Water-soluble, film-forming polysaccharide useful in coatings and films.
  • Stable solid powder with recommended refrigerated storage for long term retention.
Catalog No. 50-002-82898 Supplier Medchemexpress LLC Supplier No. HYW14552210G
Pullulan is a water-soluble linear glucosic polysaccharide produced by Aureobasidium pullulans. Supplied as a viscosity-specified powder standard for laboratory and formulation use, it offers defined molecular-weight distribution and high purity for food, pharmaceutical, and materials applications.
  • Provides controlled viscosity (10% solution: 15-180 mPa·s).
  • Defined molecular-weight distribution (Mw 362-480 kDa; Mn 100-200 kDa).
  • High purity suitable for analytical and formulation work.
  • Water-soluble, film-forming polysaccharide useful in coatings and films.
  • Stable solid powder with recommended refrigerated storage for long term retention.