Learn More
Medchemexpress LLC Pullulan | 9057-02-7 | MFCD00081940 | 98.0% | Mw 362-480 kDa; Mn 100-200 kDa | (C6H10O5)n | 25 G
SDP
Catalog No. 5000283491
Encompass_Preferred
Pullulan is a water-soluble linear glucosic polysaccharide composed of maltotriose repeating units linked by α-1,6 glycosidic bonds. It forms clear, film-forming solutions and is used as a film former, stabilizer, thickener, and excipient across food, pharmaceutical, and industrial applications. This product is supplied as a high-purity powder characterized by a 10% solution viscosity of 15-180 mPa·s.
  • Forms clear, film-forming aqueous solutions suitable for coatings and encapsulation.
  • Provides viscosity control at low concentrations (10% solution viscosity 15-180 mPa·s).
  • High purity suitable for formulation and laboratory use.
  • Biocompatible and water soluble for easy processing and removal.
  • Stable across a range of processing conditions and pH levels.
  • Applications include food coatings, pharmaceutical excipients, and industrial binders.
Catalog No. 50-002-83491 Supplier Medchemexpress LLC Supplier No. HYW14552225G
Pullulan is a water-soluble linear glucosic polysaccharide composed of maltotriose repeating units linked by α-1,6 glycosidic bonds. It forms clear, film-forming solutions and is used as a film former, stabilizer, thickener, and excipient across food, pharmaceutical, and industrial applications. This product is supplied as a high-purity powder characterized by a 10% solution viscosity of 15-180 mPa·s.
  • Forms clear, film-forming aqueous solutions suitable for coatings and encapsulation.
  • Provides viscosity control at low concentrations (10% solution viscosity 15-180 mPa·s).
  • High purity suitable for formulation and laboratory use.
  • Biocompatible and water soluble for easy processing and removal.
  • Stable across a range of processing conditions and pH levels.
  • Applications include food coatings, pharmaceutical excipients, and industrial binders.