Learn More
Malvern Panalytical Pullulan/Dextran Standrd Ls/Td
Catalog No. NC0786628
Encompass
TDS-PUL-NB - Pullulan/Dextran Standards for Light Scattering/Triple Detection calibration. Narrow calibration standard + Broad verification standard (6 sets = 12 Vials)
Catalog No. NC0786628 Supplier Malvern Panalytical Supplier No. 10021234
TDS-PUL-NB - Pullulan/Dextran Standards for Light Scattering/Triple Detection calibration. Narrow calibration standard + Broad verification standard (6 sets = 12 Vials)