Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pullulan, Spectrum™ Chemical
SDP

Catalog No. 18605954 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
50 kg
Packaging:
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-605-954 50 kg Poly Bottle
18-605-951 25 g Poly Bottle
18-605-952 100 g Poly Bottle
18-605-953 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-605-954 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. P183050KGBL
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

P1830, 9057-02-7, (C36H60O30)n

Spectrum™ Chemical Pullulan , a polysaccharide polymer, is used in the manufacture of edible films used in oral hygiene products. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Chemical Identifiers

CAS 9057-02-7
Molecular Formula (C18H30O15)AHO
SMILES *.O-*.[H][C@]1(-*)O[C@H](CO)[C@@H](O[C@@]2([H])O[C@H](CO)[C@@H](O[C@@]3([H])O[C@H](CO-*)[C@@H](O)[C@H](O)[C@H]3O)[C@H](O)[C@H]2O)[C@H](O)[C@H]1O

Specifications

CAS Max % 1
Packaging Poly Bottle
Quantity 50 kg
Grade Ungraded
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Pullulan
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.