Learn More
Pullulan Standard, Certified According to DIN, for GPC, 200,000, MilliporeSigma™ Supelco™
Catalog No. 111003859 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 mg
Catalog No. Quantity
11-100-3859 25 mg
1 options
Catalog No. 11-100-3859 Supplier MilliporeSigma Supelco Supplier No. 0161525MG
Add to Cart
Add to Cart
Pullulan Standards are mostly used to determine molecular mass distribution of cellulosic samples. It mainly consists of polymaltotriose units linked together by α-(1→6) linkages.
TRUSTED_SUSTAINABILITY
Specifications
Type Certified Reference Material
Quantity 25 mg
Grade GPC