Learn More
Creative Biomart Poractant alfa, Pulmonary surfactant
Catalog No. 502084198
Encompass_Preferred
The product is an extract of natural porcine lung surfactant consisting of 99% polar lipids (mainly phospholipids) and 1% hydrophobic low molecular weight proteins (surfactant associated proteins SP-B and SP-C). The phospholipid content of the extract consists primarily of phosphatidylcholine and dipaImitoylphosphatidylcholine. The product is a creamy white suspension of this extract in 0.9% sodium chloride solution. It contains no preservatives.
Catalog No. 50-208-4198 Supplier Creative Biomart Supplier No. THP0147
The product is an extract of natural porcine lung surfactant consisting of 99% polar lipids (mainly phospholipids) and 1% hydrophobic low molecular weight proteins (surfactant associated proteins SP-B and SP-C). The phospholipid content of the extract consists primarily of phosphatidylcholine and dipaImitoylphosphatidylcholine. The product is a creamy white suspension of this extract in 0.9% sodium chloride solution. It contains no preservatives.