Learn More
SCILUTIONS LLC PULPFIXIN 48VIAL 2ML 2D 10PK
Catalog No. NC3690883
Encompass_Preferred
PulpFixin 48 Vial, Biodegradable AutoRack for 2ml 2D Barcoded Vials - Box of 10pcs
Catalog No. NC3690883 Supplier SCILUTIONS LLC Supplier No. PFARV4802
PulpFixin 48 Vial, Biodegradable AutoRack for 2ml 2D Barcoded Vials - Box of 10pcs