Learn More
Medchemexpress LLC Pulrodemstat benzenesulfonate | 2097523-60-7 | 99.6% | 10 MG
SDP
Catalog No. 5000339421
Encompass_Preferred
Pulrodemstat benzenesulfonate (CC-90011) is a potent, selective, reversible, and orally active inhibitor of lysine specific demethylase-1 (LSD1) with an IC50 of 0.25 nM. It exhibits less enzymatic inhibition against LSD2, MAO-A, and MAO-B. This compound induces differentiation in acute myeloid leukemia (AML) and small cell lung cancer (SCLC) cells and demonstrates potent anticancer activity.
  • Potent, selective, reversible, and orally active LSD1 inhibitor
  • IC50 of 0.25 nM for LSD1
  • Induces acute myeloid leukemia (AML) and small cell lung cancer (SCLC) cell differentiation
  • Potent anticancer activity
  • Less enzymatic inhibition against LSD2, MAO-A, and MAO-B
Catalog No. 50-003-39421 Supplier Medchemexpress LLC Supplier No. HY129388B10MG
Pulrodemstat benzenesulfonate (CC-90011) is a potent, selective, reversible, and orally active inhibitor of lysine specific demethylase-1 (LSD1) with an IC50 of 0.25 nM. It exhibits less enzymatic inhibition against LSD2, MAO-A, and MAO-B. This compound induces differentiation in acute myeloid leukemia (AML) and small cell lung cancer (SCLC) cells and demonstrates potent anticancer activity.
  • Potent, selective, reversible, and orally active LSD1 inhibitor
  • IC50 of 0.25 nM for LSD1
  • Induces acute myeloid leukemia (AML) and small cell lung cancer (SCLC) cell differentiation
  • Potent anticancer activity
  • Less enzymatic inhibition against LSD2, MAO-A, and MAO-B