Learn More
Medchemexpress LLC Pulrodemstat benzenesulfonate | 2097523-60-7 | 99.6% | 50 MG
SDP
Catalog No. 5000338844
Encompass_Preferred
Pulrodemstat benzenesulfonate is a potent, selective, reversible, and orally active inhibitor of lysine specific demethylase-1 (LSD1) with an IC50 of 0.25 nM. It induces differentiation in acute myeloid leukemia (AML) and small cell lung cancer (SCLC) cells, demonstrating potent anticancer activity.
  • Potent, selective, reversible, and orally active LSD1 inhibitor
  • Less enzymatic inhibition against LSD2, MAO-A, and MAO-B
  • Induces differentiation in AML and SCLC cells
  • Demonstrates potent anticancer activity
  • Shows potent induction of CD11b in THP-1 cell line
  • Exhibits antiproliferative activity in AML kasumi-1 cells
  • Suppression of GRP observed in a dose-dependent manner
  • Potent antiproliferative activity in SCLC cells
Catalog No. 50-003-38844 Supplier Medchemexpress LLC Supplier No. HY129388B50MG
Pulrodemstat benzenesulfonate is a potent, selective, reversible, and orally active inhibitor of lysine specific demethylase-1 (LSD1) with an IC50 of 0.25 nM. It induces differentiation in acute myeloid leukemia (AML) and small cell lung cancer (SCLC) cells, demonstrating potent anticancer activity.
  • Potent, selective, reversible, and orally active LSD1 inhibitor
  • Less enzymatic inhibition against LSD2, MAO-A, and MAO-B
  • Induces differentiation in AML and SCLC cells
  • Demonstrates potent anticancer activity
  • Shows potent induction of CD11b in THP-1 cell line
  • Exhibits antiproliferative activity in AML kasumi-1 cells
  • Suppression of GRP observed in a dose-dependent manner
  • Potent antiproliferative activity in SCLC cells