Learn More
Cayman Chemical PulsatIla SaponIn D 5mg
SDP
Catalog No. 502988839
Encompass_Preferred
An oleanane-type saponin with anticancer activity; cytotoxic to A549, SKOV3, SK-MEL-2, and HCT15 cells (EC50s = 11.25, 13.17, 3.04, and 11.86 μM, respectively); reduces tumor volume in a murine Lewis lung carcinoma model at 6 mg/kg per day
Catalog No. 50-298-8839 Supplier Cayman Chemical Supplier No. 294555 MG
Add to Cart
Add to Cart
An oleanane-type saponin with anticancer activity; cytotoxic to A549, SKOV3, SK-MEL-2, and HCT15 cells (EC50s = 11.25, 13.17, 3.04, and 11.86 μM, respectively); reduces tumor volume in a murine Lewis lung carcinoma model at 6 mg/kg per day