Learn More
Medchemexpress LLC Pulsatilla saponin D | 68027-15-6 | 913.10 | 1 ML
SDP
Catalog No. 5000416980
Encompass_Preferred
Pulsatilla saponin D (SB365), also known as Hederacolchiside A, is a natural product isolated from the root of Pulsatilla chinensis. It is an anti-tumor agent.
  • Shows apoptotic effect.
  • Strongly inhibits c-Met expression in gastric cancer cells.
  • Suppresses the growth and proliferation of 5 human pancreatic cancer cell lines.
  • Exhibits anti-angiogenic activity.
  • Shows anti-tumor activity in NSCLC cancer cells.
  • Demonstrates anti-tumor activity in a Lewis lung carcinoma model.
Catalog No. 50-004-16980 Supplier Medchemexpress LLC Supplier No. HYN083410MM/1ML
Pulsatilla saponin D (SB365), also known as Hederacolchiside A, is a natural product isolated from the root of Pulsatilla chinensis. It is an anti-tumor agent.
  • Shows apoptotic effect.
  • Strongly inhibits c-Met expression in gastric cancer cells.
  • Suppresses the growth and proliferation of 5 human pancreatic cancer cell lines.
  • Exhibits anti-angiogenic activity.
  • Shows anti-tumor activity in NSCLC cancer cells.
  • Demonstrates anti-tumor activity in a Lewis lung carcinoma model.